siRNA Details
Gene Symbol: ERI3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001301698.1 | NP_001288627.1 | 3 | 728 | ||
NM_001301699.1 | NP_001288628.1 | 3 | 360 | |||
NM_001301700.1 | NP_001288629.1 | 4 | 436 | |||
NM_001301701.1 | NP_001288630.1 | 3 | 158 | |||
NM_024066.2 | NP_076971.1 | 4 | 804 | |||
XM_005271186.2 | XP_005271243.1 | 4 | 792 | |||
XM_006710891.3 | XP_006710954.1 | 3 | 461 | |||
XM_017002301.1 | XP_016857790.1 | 4 | 786 | |||
XM_017002302.1 | XP_016857791.1 | 2 | 353 | |||
XM_017002303.1 | XP_016857792.1 | 4 | 435 | |||
XM_017002304.1 | XP_016857793.1 | 2 | 420 | |||
XM_017002305.1 | XP_016857794.1 | 2 | 101 | |||
GenBank mRNA | AF007157.1 | - | 4 | 733 | ||
AK290567.1 | - | 4 | 792 | |||
AK293833.1 | - | 2 | 148 | |||
AK297708.1 | - | 2 | 350 | |||
BC001072.2 | - | 1 | 18 | |||
BC004456.1 | - | 1 | 18 | |||
BC144384.1 | - | 3 | 702 | |||
BI765887.1 | - | 3 | 432 | |||
DC413416.1 | - | 4 | 436 |

More Information
Other Species Targeted

This siRNA also targets the following transcripts in Mouse: NM_001285899, NM_001285902, NM_001285901, NM_080469, BC096545, and others.
Gene Ontology Categories:
