siRNA Details
Gene Symbol: ZFR2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_015174.1 | NP_055989.1 | 3 | 293 | ||
XM_011527831.2 | XP_011526133.1 | 3 | 292 | |||
GenBank mRNA | AB029009.1 | - | 2 | 171 | ||
AK295037.1 | - | 3 | 292 | |||
DA333602.1 | - | 3 | 293 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
