siRNA Details
Gene Symbol: ANKRD24 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_133475.1 | NP_597732.1 | 13 | 1245 | ||
XM_011527756.2 | XP_011526058.1 | 14 | 1155 | |||
XM_011527757.2 | XP_011526059.1 | 13 | 1403 | |||
XM_011527758.2 | XP_011526060.1 | 13 | 1068 | |||
XM_011527761.1 | XP_011526063.1 | 15 | 1225 | |||
XM_011527764.2 | XP_011526066.1 | 12 | 1316 | |||
XM_011527765.2 | XP_011526067.1 | 12 | 969 | |||
XM_017026438.1 | XP_016881927.1 | 15 | 1216 | |||
XM_017026439.1 | XP_016881928.1 | 14 | 1138 | |||
XM_017026440.1 | XP_016881929.1 | 13 | 1111 | |||
XM_017026441.1 | XP_016881930.1 | 3 | 253 | |||
GenBank mRNA | AB075861.1 | - | 11 | 942 | ||
BC028236.1 | - | 11 | 887 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
