siRNA Details
Gene Symbol: TBC1D9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_015130.2 | NP_055945.2 | 8 | 1663 | ||
GenBank mRNA | AB020689.2 | - | 8 | 1466 | ||
AB449892.1 | - | 8 | 1391 | |||
AK127020.1 | - | 5 | 951 | |||
BC146758.1 | - | 8 | 1466 | 9007196 | ||
CN332501.1 | - | 3 | 410 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
