siRNA Details
Gene Symbol: TBC1D9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_015130.2 | NP_055945.2 | 13 | 2560 | ||
GenBank mRNA | AB020689.2 | - | 13 | 2363 | ||
AB449892.1 | - | 13 | 2288 | |||
AK127020.1 | - | 10 | 1848 | |||
BC038804.1 | - | 4 | 674 | |||
BC146758.1 | - | 13 | 2363 | 9007196 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
