siRNA Details
Gene Symbol: ZNF264 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_003417.4 | NP_003408.1 | 4 | 839 | ||
XM_011527522.2 | XP_011525824.1 | 3 | 743 | |||
GenBank mRNA | AB007872.1 | - | 4 | 788 | ||
AK292504.1 | - | 4 | 839 | |||
BC115410.2 | - | 4 | 481 | |||
BC115411.1 | - | 5 | 521 | 40078437 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
