siRNA Details
Gene Symbol: BICC1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001080512.2 | NP_001073981.1 | 10 | 1203 | ||
XM_005270169.4 | XP_005270226.1 | 9 | 4201 | |||
XM_011540185.2 | XP_011538487.1 | 11 | 1449 | |||
XM_011540188.2 | XP_011538490.2 | 10 | 1334 | |||
XM_011540189.2 | XP_011538491.1 | 9 | 1100 | |||
XM_011540190.2 | XP_011538492.1 | 9 | 1057 | |||
XM_011540191.2 | XP_011538493.1 | 7 | 919 | |||
XM_017016677.1 | XP_016872166.1 | 11 | 7837 | |||
XM_017016678.1 | XP_016872167.1 | 12 | 1354 | |||
GenBank mRNA | AK026129.1 | - | 2 | 105 | ||
AK225427.1 | - | 2 | 105 | |||
AK225429.1 | - | 2 | 105 | |||
AK307780.1 | - | 4 | 487 | |||
AY726586.1 | - | 10 | 1148 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
