siRNA Details
Gene Symbol: BICC1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001080512.2 | NP_001073981.1 | 12, 13 | 1727 | ||
XM_005270169.4 | XP_005270226.1 | 11, 12 | 4725 | |||
XM_011540185.2 | XP_011538487.1 | 13, 14 | 1973 | |||
XM_011540188.2 | XP_011538490.2 | 12, 13 | 1858 | |||
XM_011540189.2 | XP_011538491.1 | 11, 12 | 1624 | |||
XM_011540190.2 | XP_011538492.1 | 11, 12 | 1581 | |||
XM_011540191.2 | XP_011538493.1 | 9, 10 | 1443 | |||
XM_017016677.1 | XP_016872166.1 | 13, 14 | 8361 | |||
XM_017016678.1 | XP_016872167.1 | 14, 15 | 1878 | |||
GenBank mRNA | AK026129.1 | - | 4, 5 | 629 | ||
AK225427.1 | - | 4, 5 | 629 | |||
AK225429.1 | - | 4, 5 | 629 | |||
AK307780.1 | - | 6, 7 | 1012 | |||
AY726586.1 | - | 12, 13 | 1672 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
