siRNA Details
Gene Symbol: LARP4B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_015155.2 | NP_055970.1 | 5 | 730 | ||
XM_005252432.3 | XP_005252489.1 | 5 | 739 | |||
XM_005252435.3 | XP_005252492.1 | 5 | 523 | |||
XM_011519435.2 | XP_011517737.1 | 5 | 574 | |||
XM_017015987.1 | XP_016871476.1 | 4 | 2779 | |||
XM_017015988.1 | XP_016871477.1 | 4 | 2777 | |||
XM_017015989.1 | XP_016871478.1 | 4 | 2779 | |||
XM_017015990.1 | XP_016871479.1 | 5 | 2298 | |||
XM_017015991.1 | XP_016871480.1 | 4 | 2793 | |||
XM_017015993.1 | XP_016871482.1 | 4 | 458 | |||
XM_017015994.1 | XP_016871483.1 | 5 | 495 | |||
GenBank mRNA | BC131630.1 | - | 4 | 447 | 40118844 | |
BC152443.1 | - | 4 | 454 | 8862568 | ||
D86971.2 | - | 4 | 454 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
