siRNA Details
Gene Symbol: LARP4B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_015155.2 | NP_055970.1 | 15 | 1983 | ||
XM_005252432.3 | XP_005252489.1 | 15 | 1992 | |||
XM_005252435.3 | XP_005252492.1 | 15 | 1776 | |||
XM_011519435.2 | XP_011517737.1 | 15 | 1827 | |||
XM_017015987.1 | XP_016871476.1 | 14 | 4032 | |||
XM_017015988.1 | XP_016871477.1 | 14 | 4030 | |||
XM_017015989.1 | XP_016871478.1 | 14 | 4032 | |||
XM_017015990.1 | XP_016871479.1 | 15 | 3551 | |||
XM_017015992.1 | XP_016871481.1 | 10 | 1246 | |||
XM_017015993.1 | XP_016871482.1 | 13 | 1632 | |||
XM_017015994.1 | XP_016871483.1 | 14 | 1669 | |||
GenBank mRNA | BC131630.1 | - | 14 | 1700 | 40118844 | |
BC152443.1 | - | 14 | 1707 | 8862568 | ||
CR749653.1 | - | 1 | 4157 | |||
D86971.2 | - | 14 | 1707 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
