siRNA Details
Gene Symbol: PLPPR5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001010861.2 | NP_001010861.1 | 3 | 618 | ||
NM_001037317.1 | NP_001032394.1 | 3 | 618 | |||
XM_011540836.2 | XP_011539138.1 | 3 | 612 | |||
XM_011540837.1 | XP_011539139.1 | 3 | 612 | |||
XM_011540838.2 | XP_011539140.1 | 4 | 1093 | |||
GenBank mRNA | AK295688.1 | - | 3 | 356 | ||
AY574039.1 | - | 3 | 573 | |||
AY634620.1 | - | 3 | 451 | |||
BC040174.1 | - | 3 | 355 | |||
BC136350.1 | - | 3 | 556 | 9020337 | ||
BC144000.1 | - | 3 | 556 | 9052518 | ||
DR001828.1 | - | 3 | 622 | |||
FJ472844.1 | - | 3 | 618 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
