siRNA Details
Gene Symbol: PLPPR5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001010861.2 | NP_001010861.1 | 6 | 1965 | ||
NM_001037317.1 | NP_001032394.1 | 6 | 1980 | |||
XM_011540836.2 | XP_011539138.1 | 7 | 2037 | |||
XM_011540838.2 | XP_011539140.1 | 7 | 2455 | |||
GenBank mRNA | BC040174.1 | - | 6 | 1702 | ||
BC136350.1 | - | 6 | 1903 | 9020337 | ||
BC144000.1 | - | 6 | 1915 | 9052518 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
