siRNA Details
Gene Symbol: DESI1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_015704.2 | NP_056519.1 | 6 | 821 | ||
GenBank mRNA | AF038183.1 | - | 6 | 779 | ||
BX403547.2 | - | 6 | 859 | |||
BX478162.1 | - | 1 | 167 | |||
CR456458.1 | - | 6 | 676 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
