siRNA Details
Gene Symbol: DESI1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_015704.2 | NP_056519.1 | 3 | 342 | ||
XM_005261571.4 | XP_005261628.1 | 3 | 362 | |||
GenBank mRNA | AF038183.1 | - | 3 | 300 | ||
BC093956.1 | - | 3 | 199 | 7939801 | ||
BC112179.1 | - | 3 | 199 | 8327647 | ||
BX403547.2 | - | 3 | 381 | |||
CR456458.1 | - | 3 | 197 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
