siRNA Details
Gene Symbol: CCDC61 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001267723.1 | NP_001254652.1 | 13 | 1458 | ||
XM_005259191.3 | XP_005259248.1 | 13 | 3563 | |||
XM_005259192.4 | XP_005259249.1 | 13 | 2204 | |||
XM_011527257.2 | XP_011525559.1 | 13 | 1487 | |||
GenBank mRNA | CA775840.1 | - | 5 | 396 | ||
CB852675.1 | - | 6 | 445 | |||
GQ375050.1 | - | 9 | 891 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
