siRNA Details
Gene Symbol: CCDC61 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001267723.1 | NP_001254652.1 | 3, 4 | 290 | ||
XM_005259191.3 | XP_005259248.1 | 3, 4 | 2395 | |||
XM_005259192.4 | XP_005259249.1 | 3, 4 | 1036 | |||
XM_011527257.2 | XP_011525559.1 | 3, 4 | 319 | |||
GenBank mRNA | DR007040.1 | - | 3, 4 | 310 | ||
GQ375050.1 | - | 3, 4 | 263 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
