siRNA Details
Gene Symbol: USP35 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_020798.2 | NP_065849.1 | 4 | 1266 | ||
XM_011545187.2 | XP_011543489.1 | 4 | 409 | |||
XM_011545188.2 | XP_011543490.1 | 2 | 192 | |||
XM_017018052.1 | XP_016873541.1 | 4 | 1263 | |||
XM_017018053.1 | XP_016873542.1 | 3 | 258 | |||
GenBank mRNA | AB037793.1 | - | 1 | 729 | ||
AJ586137.1 | - | 3 | 907 | |||
AK307875.1 | - | 3 | 492 | |||
DB095282.1 | - | 3 | 522 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
