siRNA Details
Gene Symbol: USP35 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_020798.2 | NP_065849.1 | 7 | 1677 | ||
XM_011545187.2 | XP_011543489.1 | 7 | 820 | |||
XM_011545188.2 | XP_011543490.1 | 5 | 603 | |||
XM_017018052.1 | XP_016873541.1 | 7 | 1674 | |||
XM_017018053.1 | XP_016873542.1 | 6 | 669 | |||
GenBank mRNA | AB037793.1 | - | 4 | 1140 | ||
AJ586137.1 | - | 6 | 1318 | |||
AK056231.1 | - | 2 | 470 | |||
AK307875.1 | - | 6 | 903 | |||
BC131489.1 | - | 5 | 640 | 40038029 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
