siRNA Details
Gene Symbol: WIZ | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_021241.2 | NP_067064.2 | 8 | 3441 | ||
XM_005260004.4 | XP_005260061.1 | 13 | 6676 | |||
XM_005260005.4 | XP_005260062.1 | 12 | 6505 | |||
XM_005260006.4 | XP_005260063.1 | 12 | 6277 | |||
XM_005260007.4 | XP_005260064.1 | 11 | 6106 | |||
XM_005260008.2 | XP_005260065.1 | 11 | 4520 | |||
XM_005260009.2 | XP_005260066.1 | 9 | 3867 | |||
XM_005260010.3 | XP_005260067.1 | 9 | 3880 | |||
XM_005260011.2 | XP_005260068.1 | 9 | 3950 | |||
XM_005260012.2 | XP_005260069.1 | 8 | 3468 | |||
XM_006722828.3 | XP_006722891.1 | 11 | 4289 | |||
XM_011528163.2 | XP_011526465.1 | 9 | 3941 | |||
GenBank mRNA | AK027615.1 | - | 5 | 2395 | ||
AK122890.1 | - | 9 | 3497 | |||
AK131404.1 | - | 8 | 3442 | |||
AL390184.1 | - | 3 | 1313 | |||
BC002329.2 | - | 3 | 1304 | |||
BC062360.1 | - | 5 | 2165 | |||
BC098445.1 | - | 5 | 2675 |

More Information
Other Species Targeted

This siRNA also targets the following transcripts in Mouse: NM_212438, NM_011717, CA325706, AB012266, AK004274, and others.
Gene Ontology Categories:
