siRNA Details
Gene Symbol: WIZ | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_021241.2 | NP_067064.2 | 8 | 3543 | ||
XM_005260004.4 | XP_005260061.1 | 13 | 6778 | |||
XM_005260005.4 | XP_005260062.1 | 12 | 6607 | |||
XM_005260006.4 | XP_005260063.1 | 12 | 6379 | |||
XM_005260007.4 | XP_005260064.1 | 11 | 6208 | |||
XM_005260008.2 | XP_005260065.1 | 11 | 4622 | |||
XM_005260009.2 | XP_005260066.1 | 9 | 3969 | |||
XM_005260010.3 | XP_005260067.1 | 9 | 3982 | |||
XM_005260011.2 | XP_005260068.1 | 9 | 4052 | |||
XM_005260012.2 | XP_005260069.1 | 8 | 3570 | |||
XM_006722828.3 | XP_006722891.1 | 11 | 4391 | |||
XM_011528163.2 | XP_011526465.1 | 9 | 4043 | |||
GenBank mRNA | AK027615.1 | - | 5 | 2497 | ||
AK122890.1 | - | 9 | 3599 | |||
AK131404.1 | - | 8 | 3544 | |||
AL390184.1 | - | 3 | 1415 | |||
BC002329.2 | - | 3 | 1406 | |||
BC062360.1 | - | 5 | 2267 | |||
BC098445.1 | - | 5 | 2777 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
