siRNA Details
Gene Symbol: DNAJC6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001256864.1 | NP_001243793.1 | 4 | 666 | ||
NM_001256865.1 | NP_001243794.1 | 5 | 614 | |||
NM_014787.3 | NP_055602.1 | 4 | 501 | |||
GenBank mRNA | AB007942.1 | - | 4 | 441 | ||
AK296408.1 | - | 5 | 568 | |||
BC051722.1 | - | 4 | 450 | |||
BC109279.2 | - | 4 | 599 | 40028812 | ||
BC109280.1 | - | 4 | 602 | 40028814 | ||
BX647203.1 | - | 4 | 655 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
