siRNA Details
Gene Symbol: CEP126 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_020802.3 | NP_065853.3 | 6 | 1681 | ||
XM_005271627.4 | XP_005271684.2 | 6 | 1412 | |||
GenBank mRNA | AB037798.1 | - | 3 | 1024 | ||
AK095004.1 | - | 5 | 1311 | |||
AK296350.1 | - | 3 | 1422 | |||
BX647415.1 | - | 6 | 1431 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
