siRNA Details
Gene Symbol: LTK | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001135685.1 | NP_001129157.1 | 13 | 1701 | ||
NM_002344.5 | NP_002335.2 | 15 | 2091 | |||
NM_206961.3 | NP_996844.1 | 14 | 1908 | |||
XM_011521556.2 | XP_011519858.1 | 15 | 2139 | |||
XM_011521557.2 | XP_011519859.1 | 14 | 1971 | |||
XM_017022181.1 | XP_016877670.1 | 15 | 2154 | |||
XM_017022182.1 | XP_016877671.1 | 14 | 2010 | |||
XM_017022183.1 | XP_016877672.1 | 14 | 1899 | |||
GenBank mRNA | AK296892.1 | - | 13 | 1701 | ||
AK314006.1 | - | 15 | 2058 | |||
BC045607.1 | - | 14 | 2335 | 5270700 | ||
BC143903.1 | - | 9 | 1094 | |||
D16105.1 | - | 15 | 2010 | |||
X52213.1 | - | 8 | 974 | |||
X60702.1 | - | 14 | 1900 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
