siRNA Details
Gene Symbol: CEP126 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_020802.3 | NP_065853.3 | 4 | 712 | ||
XM_005271627.4 | XP_005271684.2 | 4 | 443 | |||
GenBank mRNA | AB037798.1 | - | 1 | 55 | ||
AK310418.1 | - | 2 | 84 | |||
BX647415.1 | - | 4 | 462 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
