siRNA Details
Gene Symbol: KCNT1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001272003.1 | NP_001258932.1 | 6 | 510 | ||
NM_020822.2 | NP_065873.2 | 7 | 654 | |||
XM_011518877.2 | XP_011517179.1 | 7 | 4624 | |||
XM_011518878.2 | XP_011517180.1 | 7 | 4624 | |||
XM_011518879.2 | XP_011517181.1 | 7 | 4624 | |||
XM_011518880.1 | XP_011517182.1 | 6 | 481 | |||
XM_011518881.2 | XP_011517183.1 | 4 | 1532 | |||
XM_017014931.1 | XP_016870420.1 | 5 | 520 | |||
XM_017014932.1 | XP_016870421.1 | 6 | 412 | |||
XM_017014933.1 | XP_016870422.1 | 4 | 239 | |||
GenBank mRNA | AB037843.1 | - | 7 | 647 | ||
AK127272.1 | - | 6 | 510 | |||
BC136618.1 | - | 7 | 626 | 9020608 | ||
BC171770.1 | - | 7 | 626 | 9054424 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
