siRNA Details
Gene Symbol: CNTN3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_020872.2 | NP_065923.1 | 17 | 2387 | ||
XM_005264757.3 | XP_005264814.1 | 18 | 2497 | |||
XM_011533768.2 | XP_011532070.1 | 17 | 6019 | |||
XM_017006507.1 | XP_016861996.1 | 18 | 3067 | |||
XM_017006508.1 | XP_016861997.1 | 17 | 2292 | |||
XM_017006509.1 | XP_016861998.1 | 8 | 1066 | |||
GenBank mRNA | AB040929.1 | - | 15 | 2036 | ||
BC150608.1 | - | 17 | 2387 | 9056978 | ||
BX951442.1 | - | 1 | 40 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
