siRNA Details
Gene Symbol: SPCS2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_014752.2 | NP_055567.2 | 5 | 893 | ||
GenBank mRNA | AK293128.1 | - | 5 | 874 | ||
BC008063.2 | - | 1 | 754 | 2967488 | ||
BC070276.1 | - | 1 | 631 | 4656138 | ||
BC082231.1 | - | 1 | 748 | 3954096 | ||
BC106066.1 | - | 5 | 858 | 4521501 | ||
D14658.1 | - | 5 | 852 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
