siRNA Details
Gene Symbol: TTC7A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001288951.1 | NP_001275880.1 | 8, 9 | 1440 | ||
NM_001288953.1 | NP_001275882.1 | 9, 10 | 1431 | |||
NM_001288955.1 | NP_001275884.1 | 7, 8 | 1282 | |||
NM_020458.3 | NP_065191.2 | 8, 9 | 1440 | |||
XM_005264439.3 | XP_005264496.1 | 9, 10 | 1593 | |||
XM_011532998.2 | XP_011531300.1 | 10, 11 | 1658 | |||
XM_011532999.2 | XP_011531301.1 | 8, 9 | 1383 | |||
XM_011533000.2 | XP_011531302.1 | 5, 6 | 515 | |||
XM_017004524.1 | XP_016860013.1 | 8, 9 | 1384 | |||
XM_017004525.1 | XP_016860014.1 | 8, 9 | 925 | |||
XM_017004526.1 | XP_016860015.1 | 8, 9 | 1384 | |||
XM_017004527.1 | XP_016860016.1 | 8, 9 | 1535 | |||
XM_017004528.1 | XP_016860017.1 | 9, 10 | 1600 | |||
XM_017004529.1 | XP_016860018.1 | 8, 9 | 1383 | |||
XM_017004530.1 | XP_016860019.1 | 8, 9 | 1384 | |||
GenBank mRNA | AB032966.2 | - | 9, 10 | 1020 | ||
AK056464.1 | - | 9, 10 | 1403 | |||
AL834383.1 | - | 8, 9 | 1440 | |||
BC065554.1 | - | 5, 6 | 558 | |||
BC082965.1 | - | 2, 3 | 355 | |||
BC111487.1 | - | 8, 9 | 1328 | 6041916 | ||
BC114365.2 | - | 7, 8 | 913 | 40038218 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
