siRNA Details
Gene Symbol: GPR108 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001080452.1 | NP_001073921.1 | 15 | 1379 | ||
XM_006722801.3 | XP_006722864.1 | 15 | 1543 | |||
XM_011528137.2 | XP_011526439.1 | 15 | 1325 | |||
XM_017027012.1 | XP_016882501.1 | 15 | 1337 | |||
XM_017027013.1 | XP_016882502.1 | 15 | 1337 | |||
XM_017027014.1 | XP_016882503.1 | 15 | 1337 | |||
XM_017027015.1 | XP_016882504.1 | 15 | 1525 | |||
GenBank mRNA | AK291968.1 | - | 15 | 1337 | ||
AL365404.1 | - | 15 | 1312 | |||
AX775899.1 | - | 15 | 1360 | |||
BC007862.2 | - | 2 | 63 | |||
BC146909.1 | - | 15 | 1353 | 9056837 | ||
BC150657.1 | - | 14 | 1157 | 9057028 | ||
EB387948.1 | - | 9 | 748 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Set Membership: |
![]() |
