siRNA Details
Gene Symbol: POGZ | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001194937.1 | NP_001181866.1 | 12 | 2174 | ||
NM_001194938.1 | NP_001181867.1 | 11 | 2015 | |||
NM_015100.3 | NP_055915.2 | 12 | 2201 | |||
NM_145796.3 | NP_665739.3 | 10 | 1916 | |||
NM_207171.2 | NP_997054.1 | 10 | 1728 | |||
XM_005244999.2 | XP_005245056.1 | 12 | 4869 | |||
XM_005245000.4 | XP_005245057.1 | 12 | 2760 | |||
XM_005245001.2 | XP_005245058.1 | 12 | 1985 | |||
XM_005245005.1 | XP_005245062.1 | 11 | 1764 | |||
XM_005245006.4 | XP_005245063.1 | 11 | 2680 | |||
XM_011509331.2 | XP_011507633.1 | 10 | 1777 | |||
XM_017000744.1 | XP_016856233.1 | 12 | 2003 | |||
XM_017000745.1 | XP_016856234.1 | 12 | 4843 | |||
XM_017000746.1 | XP_016856235.1 | 12 | 2734 | |||
XM_017000748.1 | XP_016856237.1 | 11 | 1747 | |||
XM_017000749.1 | XP_016856238.1 | 11 | 2118 | |||
GenBank mRNA | AB007930.1 | - | 10 | 1723 | ||
AB037911.1 | - | 11 | 1783 | |||
AB075477.1 | - | 11 | 1783 | |||
AJ242979.1 | - | 12 | 2192 | |||
AK300307.1 | - | 12 | 1888 | |||
AK302501.1 | - | 11 | 1765 | |||
BX537561.1 | - | 10 | 1630 | |||
BX537838.1 | - | 12 | 2202 | |||
BX649163.1 | - | 11 | 1756 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
