siRNA Details
Gene Symbol: HEATR1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_018072.5 | NP_060542.4 | 9 | 1290 | ||
GenBank mRNA | AK095692.1 | - | 9 | 1253 | ||
BC150614.1 | - | 8 | 1160 | 9056984 | ||
BU102172.1 | - | 4 | 431 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
