siRNA Details
Gene Symbol: KIAA1211 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_020722.1 | NP_065773.1 | 8 | 2615 | ||
XM_005265752.2 | XP_005265809.1 | 7 | 2622 | |||
XM_005265755.2 | XP_005265812.1 | 4 | 2051 | |||
XM_005265756.2 | XP_005265813.1 | 4 | 2099 | |||
XM_006714045.2 | XP_006714108.1 | 4 | 2076 | |||
XM_011534397.2 | XP_011532699.1 | 7 | 3365 | |||
XM_017008473.1 | XP_016863962.1 | 7 | 2351 | |||
XM_017008474.1 | XP_016863963.1 | 6 | 3229 | |||
GenBank mRNA | AB033037.1 | - | 2 | 936 | ||
AK097530.1 | - | 5 | 1145 | |||
AK126014.1 | - | 8 | 2615 | |||
AK302486.1 | - | 4 | 2117 | |||
AL133028.1 | - | 1 | 1027 | |||
AL137336.1 | - | 1 | 393 | |||
BC144536.1 | - | 7 | 2369 | 9053063 | ||
BC150615.1 | - | 7 | 2369 | 9056985 | ||
BC171783.1 | - | 7 | 2351 | 9054437 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
