siRNA Details
Gene Symbol: SLAIN2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_020846.1 | NP_065897.1 | 6 | 1422 | ||
XM_005248121.2 | XP_005248178.1 | 6 | 1538 | |||
GenBank mRNA | AB040891.1 | - | 6 | 1392 | ||
AK025264.1 | - | 2 | 473 | |||
AK026476.1 | - | 6 | 969 | |||
AK291006.1 | - | 6 | 1552 | |||
BC031691.2 | - | 4 | 742 | |||
BC040993.1 | - | 6 | 1701 | 5272534 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
