siRNA Details
Gene Symbol: SLAIN2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_020846.1 | NP_065897.1 | 8 | 2104 | ||
XM_005248121.2 | XP_005248178.1 | 9 | 2298 | |||
GenBank mRNA | AB040891.1 | - | 8 | 2074 | ||
AK026476.1 | - | 8 | 1652 | |||
AK291006.1 | - | 8 | 2234 | |||
BC031691.2 | - | 6 | 1424 | |||
BC040993.1 | - | 8 | 2383 | 5272534 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
