siRNA Details
Gene Symbol: FRYL | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_015030.1 | NP_055845.1 | 43 | 5934 | ||
XM_005248090.4 | XP_005248147.1 | 44 | 5992 | |||
XM_005248093.4 | XP_005248150.1 | 18 | 2619 | |||
XM_011513680.2 | XP_011511982.1 | 41 | 5451 | |||
XM_011513681.2 | XP_011511983.1 | 41 | 5454 | |||
XM_011513682.2 | XP_011511984.1 | 40 | 5409 | |||
XM_011513683.2 | XP_011511985.1 | 43 | 5828 | |||
XM_011513684.2 | XP_011511986.1 | 18 | 2625 | |||
XM_017008046.1 | XP_016863535.1 | 44 | 6259 | |||
GenBank mRNA | AK127307.1 | - | 5 | 679 | ||
AL833170.1 | - | 10 | 1455 | |||
U80082.1 | - | 2 | 742 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
