siRNA Details
Gene Symbol: FNIP2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001323916.1 | NP_001310845.1 | 2 | 410 | ||
NM_020840.2 | NP_065891.1 | 2 | 405 | |||
XM_005263156.1 | XP_005263213.1 | 2 | 405 | |||
XM_005263158.1 | XP_005263215.1 | 2 | 205 | |||
XM_005263160.3 | XP_005263217.1 | 2 | 405 | |||
XM_011532154.2 | XP_011530456.1 | 2 | 1185 | |||
XM_017008487.1 | XP_016863976.1 | 2 | 405 | |||
GenBank mRNA | AB040883.1 | - | 2 | 262 | ||
AL713802.1 | - | 2 | 206 | |||
BC016638.1 | - | 2 | 410 | |||
DB482145.1 | - | 2 | 371 | |||
HY049314.1 | - | 2 | 407 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
