siRNA Details
Gene Symbol: FNIP2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001323916.1 | NP_001310845.1 | 10 | 1298 | ||
NM_020840.2 | NP_065891.1 | 10 | 1293 | |||
XM_005263156.1 | XP_005263213.1 | 11 | 1383 | |||
XM_005263158.1 | XP_005263215.1 | 11 | 1183 | |||
XM_005263160.3 | XP_005263217.1 | 11 | 1383 | |||
XM_011532154.2 | XP_011530456.1 | 11 | 2163 | |||
XM_017008487.1 | XP_016863976.1 | 12 | 1332 | |||
GenBank mRNA | AB040883.1 | - | 10 | 1150 | ||
BC016638.1 | - | 10 | 1298 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
