siRNA Details
Gene Symbol: FNIP2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001323916.1 | NP_001310845.1 | 16 | 3478 | ||
NM_020840.2 | NP_065891.1 | 16 | 3473 | |||
XM_005263156.1 | XP_005263213.1 | 17 | 3563 | |||
XM_005263158.1 | XP_005263215.1 | 17 | 3363 | |||
XM_011532154.2 | XP_011530456.1 | 17 | 4343 | |||
XM_017008487.1 | XP_016863976.1 | 18 | 3512 | |||
GenBank mRNA | AB040883.1 | - | 16 | 3330 | ||
AK022968.1 | - | 4 | 1270 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
