siRNA Details
Gene Symbol: FRMD4B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_015123.2 | NP_055938.1 | 8 | 942 | ||
XM_005264722.1 | XP_005264779.1 | 8 | 619 | |||
XM_017005989.1 | XP_016861478.1 | 8 | 726 | |||
XM_017005990.1 | XP_016861479.1 | 8 | 619 | |||
XM_017005991.1 | XP_016861480.1 | 9 | 779 | |||
XM_017005992.1 | XP_016861481.1 | 8 | 559 | |||
XM_017005993.1 | XP_016861482.1 | 8 | 550 | |||
XM_017005994.1 | XP_016861483.1 | 8 | 2392 | |||
XM_017005995.1 | XP_016861484.1 | 2 | 136 | |||
GenBank mRNA | AB023230.1 | - | 8 | 711 | ||
AK000244.1 | - | 8 | 523 | |||
AK295045.1 | - | 3 | 184 | |||
DC340455.1 | - | 3 | 153 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
