siRNA Details
Gene Symbol: SHC2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_012435.2 | NP_036567.2 | 10, 11 | 1301 | ||
XM_011527893.2 | XP_011526195.1 | 10, 11 | 1316 | |||
XM_011527896.2 | XP_011526198.1 | 12, 13 | 1361 | |||
XM_017026567.1 | XP_016882056.1 | 11, 12 | 1449 | |||
GenBank mRNA | AB001451.1 | - | 10, 11 | 1175 | ||
AL360254.1 | - | 8, 9 | 731 | |||
BY796046.2 | - | 1, 2 | 118 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
