siRNA Details
Gene Symbol: SHC2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_012435.2 | NP_036567.2 | 5, 6 | 781 | ||
XM_011527893.2 | XP_011526195.1 | 5, 6 | 796 | |||
XM_011527894.2 | XP_011526196.1 | 5, 6 | 796 | |||
XM_011527895.2 | XP_011526197.1 | 5, 6 | 796 | |||
XM_011527896.2 | XP_011526198.1 | 7, 8 | 841 | |||
XM_017026567.1 | XP_016882056.1 | 6, 7 | 929 | |||
GenBank mRNA | AB001451.1 | - | 5, 6 | 655 | ||
AL134433.1 | - | 5, 6 | 438 | |||
AL360254.1 | - | 3, 4 | 211 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
