siRNA Details
Gene Symbol: SHC2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_012435.2 | NP_036567.2 | 12 | 1636 | ||
XM_011527895.2 | XP_011526197.1 | 11 | 1516 | |||
GenBank mRNA | AB001451.1 | - | 12 | 1510 | ||
AL360254.1 | - | 10 | 1066 | |||
BC048290.1 | - | 2 | 338 | |||
BY796046.2 | - | 3 | 453 | |||
CF457169.1 | - | 3 | 438 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
