siRNA Details
Gene Symbol: PHRF1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001286581.1 | NP_001273510.1 | 5 | 615 | ||
NM_001286582.1 | NP_001273511.1 | 5 | 612 | |||
NM_001286583.1 | NP_001273512.1 | 5 | 603 | |||
NM_020901.3 | NP_065952.2 | 5 | 615 | |||
XM_005253025.4 | XP_005253082.1 | 5 | 573 | |||
XM_005253027.3 | XP_005253084.1 | 5 | 561 | |||
XM_011520236.2 | XP_011518538.1 | 5 | 615 | |||
XM_011520237.2 | XP_011518539.1 | 4 | 456 | |||
GenBank mRNA | AB040975.1 | - | 5 | 575 | ||
BC136615.1 | - | 5 | 540 | 9020605 | ||
BC144293.1 | - | 5 | 540 | 9052813 | ||
BC144294.1 | - | 5 | 528 | 9052814 | ||
BC144295.1 | - | 5 | 537 | 9052815 | ||
BC146771.1 | - | 5 | 575 | 9007218 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
