siRNA Details
Gene Symbol: PHRF1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001286581.1 | NP_001273510.1 | 4 | 575 | ||
NM_001286582.1 | NP_001273511.1 | 4 | 572 | |||
NM_001286583.1 | NP_001273512.1 | 4 | 563 | |||
NM_020901.3 | NP_065952.2 | 4 | 575 | |||
XM_005253025.4 | XP_005253082.1 | 4 | 533 | |||
XM_005253027.3 | XP_005253084.1 | 4 | 521 | |||
XM_011520236.2 | XP_011518538.1 | 4 | 575 | |||
XM_011520237.2 | XP_011518539.1 | 3 | 416 | |||
GenBank mRNA | AB040975.1 | - | 4 | 535 | ||
BC112931.1 | - | 3 | 846 | |||
BC136615.1 | - | 4 | 500 | 9020605 | ||
BC144293.1 | - | 4 | 500 | 9052813 | ||
BC144294.1 | - | 4 | 488 | 9052814 | ||
BC144295.1 | - | 4 | 497 | 9052815 | ||
BC146771.1 | - | 4 | 535 | 9007218 | ||
DA989169.1 | - | 4 | 538 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
