siRNA Details
Gene Symbol: PHRF1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001286581.1 | NP_001273510.1 | 18 | 5038 | ||
NM_001286582.1 | NP_001273511.1 | 18 | 5032 | |||
NM_001286583.1 | NP_001273512.1 | 18 | 5026 | |||
NM_020901.3 | NP_065952.2 | 18 | 5035 | |||
XM_005253025.4 | XP_005253082.1 | 18 | 4996 | |||
XM_005253027.3 | XP_005253084.1 | 18 | 4984 | |||
XM_011520236.2 | XP_011518538.1 | 18 | 5032 | |||
XM_011520237.2 | XP_011518539.1 | 17 | 4879 | |||
GenBank mRNA | AB040975.1 | - | 18 | 4998 | ||
BC004950.1 | - | 5 | 890 | |||
BC013381.2 | - | 5 | 1514 | |||
BC029651.1 | - | 5 | 3164 | |||
BC041631.1 | - | 5 | 1425 | |||
BC112931.1 | - | 16 | 6768 | |||
BC127876.1 | - | 2 | 224 | |||
BC136615.1 | - | 18 | 4963 | 9020605 | ||
BC144293.1 | - | 18 | 4960 | 9052813 | ||
BC144294.1 | - | 18 | 4951 | 9052814 | ||
BC144295.1 | - | 18 | 4957 | 9052815 | ||
BC146771.1 | - | 18 | 4998 | 9007218 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
