siRNA Details
Gene Symbol: EFR3B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001319099.1 | NP_001306028.1 | 5 | 670 | ||
NM_014971.1 | NP_055786.1 | 5 | 682 | |||
XM_011532701.2 | XP_011531003.1 | 5 | 682 | |||
GenBank mRNA | AB023170.1 | - | 5 | 656 | ||
AK095190.1 | - | 5 | 670 | |||
BC049384.1 | - | 2 | 226 | 5742159 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
