siRNA Details
Gene Symbol: WDR1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_017491.3 | NP_059830.1 | 4 | 608 | ||
XM_017008880.1 | XP_016864369.1 | 4 | 642 | |||
GenBank mRNA | AB010427.2 | - | 4 | 527 | ||
AB209746.1 | - | 4 | 381 | |||
AF020056.1 | - | 4 | 436 | |||
AF020260.1 | - | 4 | 436 | |||
AF274954.1 | - | 362 | ||||
AK222764.1 | - | 4 | 480 | |||
AK222897.1 | - | 4 | 512 | |||
AK225276.1 | - | 4 | 480 | |||
AK291614.1 | - | 4 | 458 | |||
AL050108.1 | - | 2 | 156 | |||
BC000201.1 | - | 4 | 407 | 3351638 | ||
BC002489.2 | - | 4 | 399 | 3051319 | ||
EU794675.1 | - | 4 | 399 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
