siRNA Details
Gene Symbol: WDR1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_017491.3 | NP_059830.1 | 5 | 792 | ||
XM_017008880.1 | XP_016864369.1 | 5 | 826 | |||
GenBank mRNA | AB010427.2 | - | 5 | 711 | ||
AB209746.1 | - | 5 | 565 | |||
AF020056.1 | - | 5 | 620 | |||
AF020260.1 | - | 5 | 620 | |||
AK127665.1 | - | 5 | 532 | |||
AK222764.1 | - | 5 | 664 | |||
AK222897.1 | - | 5 | 696 | |||
AK225276.1 | - | 5 | 664 | |||
AK291614.1 | - | 5 | 642 | |||
AK299602.1 | - | 5 | 468 | |||
AL050108.1 | - | 3 | 340 | |||
BC000201.1 | - | 5 | 591 | 3351638 | ||
BC002489.2 | - | 5 | 583 | 3051319 | ||
EU794675.1 | - | 5 | 583 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
