siRNA Details
Gene Symbol: FAM184B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_015688.1 | NP_056503.1 | 18 | 3374 | ||
GenBank mRNA | AB033102.1 | - | 19 | 3187 | ||
AK027311.1 | - | 2 | 285 | |||
AK308144.1 | - | 19 | 3375 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
