siRNA Details
Gene Symbol: KIAA1257 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_020741.2 | NP_065792.1 | 6 | 1083 | ||
XM_011513050.2 | XP_011511352.2 | 6 | 983 | |||
XM_017006939.1 | XP_016862428.1 | 7 | 1538 | |||
XM_017006940.1 | XP_016862429.1 | 7 | 1534 | |||
XM_017006941.1 | XP_016862430.1 | 7 | 1111 | |||
XM_017006942.1 | XP_016862431.1 | 7 | 1505 | |||
XM_017006943.1 | XP_016862432.1 | 7 | 1263 | |||
XM_017006944.1 | XP_016862433.1 | 6 | 1083 | |||
XM_017006945.1 | XP_016862434.1 | 6 | 983 | |||
XM_017006946.1 | XP_016862435.1 | 5 | 694 | |||
XM_017006947.1 | XP_016862436.1 | 4 | 578 | |||
XM_017006948.1 | XP_016862437.1 | 6 | 755 | |||
XM_017006949.1 | XP_016862438.1 | 5 | 702 | |||
XM_017006950.1 | XP_016862439.1 | 5 | 6189 | |||
XM_017006951.1 | XP_016862440.1 | 5 | 6294 | |||
XM_017006952.1 | XP_016862441.1 | 4 | 472 | |||
XM_017006953.1 | XP_016862442.1 | 4 | 478 | |||
GenBank mRNA | AB033083.1 | - | 6 | 1055 | ||
BC031632.1 | - | 5 | 1044 | 5170293 | ||
BC038736.1 | - | 6 | 1016 |
Gene Symbol: LOC100132731 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK056377.1 | - | 5 | 1659 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
